Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300010650.mm.1 |
NR_045053.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010650.mm.1 |
NR_045053.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010650.mm.1 |
NR_045054.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010650.mm.1 |
NR_045054.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010650.mm.1 |
TC1300001707.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010649.mm.1 |
NR_045053.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010649.mm.1 |
NR_045053.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010649.mm.1 |
NR_045054.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010649.mm.1 |
NR_045054.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010649.mm.1 |
TC1300001707.mm.1 |
MTA TC |