Assay Details
| Gene Symbol: Rsbn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080977.1 | NP_001074446.1 | 4 - 5 | 1454 | 118 | |
| GenBank mRNA | AK033374.1 | - | 4 - 5 | 1134 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rsbn1l
Genomic Map