Assay Details
Gene Symbol: Rsbn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080977.1 | NP_001074446.1 | 5 - 6 | 1598 | 77 | |
GenBank mRNA | AK033374.1 | - | 5 - 6 | 1278 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
