Assay Details
| Gene Symbol: Slc4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253892.1 | NP_001240821.1 | 12 - 13 | 2038 | 122 | |
| NM_009207.3 | NP_033233.2 | 13 - 14 | 2234 | 122 | ||
| GenBank mRNA | AK145930.1 | - | 13 - 14 | 2159 | 122 | |
| AK147858.1 | - | 13 - 14 | 2190 | 122 | ||
| BC054102.1 | - | 12 - 13 | 2043 | 30017310 | 122 | |
| J04036.1 | - | 13 - 14 | 2149 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a2
Genomic Map