Assay Details
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007462.3 | NP_031488.2 | 2 - 3 | 450 | 95 | |
GenBank mRNA | AK042021.1 | - | 2 - 3 | 451 | 95 | |
AK045053.1 | - | 2 - 3 | 421 | 95 | ||
AK046280.1 | - | 2 - 3 | 441 | 95 | ||
AK080907.1 | - | 2 - 3 | 450 | 95 | ||
AK082724.1 | - | 2 - 3 | 196 | 95 | ||
BC141141.1 | - | 1 - 2 | 163 | 9055753 | 95 | |
M88127.1 | - | 1 - 2 | 139 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
