Assay Details
| Gene Symbol: Zc3hav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028421.1 | NP_082697.1 | 3 - 4 | 1075 | 65 | |
| NM_028864.2 | NP_083140.1 | 3 - 4 | 1075 | 65 | ||
| GenBank mRNA | AK004770.1 | - | 3 - 4 | 1076 | 65 | |
| AK020263.2 | - | 3 - 4 | 754 | 65 | ||
| AK080334.1 | - | 3 - 4 | 1060 | 65 | ||
| AK143568.1 | - | 3 - 4 | 745 | 65 | ||
| AK169402.1 | - | 3 - 4 | 720 | 65 | ||
| AK172379.1 | - | 3 - 4 | 1076 | 65 | ||
| BC029090.1 | - | 3 - 4 | 985 | 2651558 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zc3hav1
Genomic Map