Assay Details
Gene Symbol: Rnf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025999.3 | NP_080275.2 | 3 - 4 | 351 | 63 | |
GenBank mRNA | AF353167.1 | - | 3 - 4 | 381 | 63 | |
AK006791.1 | - | 3 - 4 | 391 | 63 | ||
AK011839.1 | - | 3 - 4 | 340 | 63 | ||
AK015505.1 | - | 3 - 4 | 352 | 63 | ||
AK139910.1 | - | 3 - 4 | 345 | 63 | ||
AK154417.1 | - | 3 - 4 | 375 | 63 | ||
BC018553.1 | - | 3 - 4 | 345 | 3582546 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
