Assay Details
| Gene Symbol: Cdc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178347.4 | NP_848124.1 | 12 - 13 | 1390 | 90 | |
| GenBank mRNA | AK031459.1 | - | 12 - 13 | 1373 | 90 | |
| AK038523.1 | - | 12 - 13 | 1373 | 90 | ||
| AK047789.1 | - | 12 - 13 | 1373 | 90 | ||
| AK048246.1 | - | 12 - 13 | 1373 | 90 | ||
| AK052567.1 | - | 12 - 13 | 1390 | 90 | ||
| AK075948.1 | - | 12 - 13 | 1391 | 90 | ||
| AK079681.1 | - | 10 - 11 | 1738 | 90 | ||
| AK081461.1 | - | 12 - 13 | 1391 | 90 | ||
| AK169677.1 | - | 8 - 9 | 864 | 90 | ||
| BC059013.1 | - | 12 - 13 | 1359 | 6813730 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc23
Genomic Map