Assay Details
| Gene Symbol: Cdc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178347.4 | NP_848124.1 | 1 - 2 | 192 | 75 | |
| GenBank mRNA | AK031459.1 | - | 1 - 2 | 175 | 75 | |
| AK038523.1 | - | 1 - 2 | 175 | 75 | ||
| AK044478.1 | - | 1 - 2 | 181 | 75 | ||
| AK047789.1 | - | 1 - 2 | 175 | 75 | ||
| AK048246.1 | - | 1 - 2 | 175 | 75 | ||
| AK052567.1 | - | 1 - 2 | 192 | 75 | ||
| AK075948.1 | - | 1 - 2 | 193 | 75 | ||
| AK081461.1 | - | 1 - 2 | 193 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc23
Genomic Map