Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000010713.mm.1 |
NM_001110218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010713.mm.1 |
NM_001110218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010713.mm.1 |
NM_176919.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010713.mm.1 |
NM_176919.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010713.mm.1 |
TC1000001512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010717.mm.1 |
NM_001110218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010717.mm.1 |
NM_001110218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010717.mm.1 |
NM_176919.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010717.mm.1 |
NM_176919.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010717.mm.1 |
TC1000001512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010714.mm.1 |
NM_001110218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010714.mm.1 |
NM_001110218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010714.mm.1 |
NM_176919.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010714.mm.1 |
NM_176919.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010714.mm.1 |
NR_106182.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010714.mm.1 |
TC1000001512.mm.1 |
MTA TC |