Assay Details
| Gene Symbol: Rsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081267.2 | NP_001074736.1 | 2 - 3 | 292 | 73 | |
| GenBank mRNA | AK032853.1 | - | 2 - 3 | 268 | 73 | |
| AK146154.1 | - | 2 - 3 | 296 | 73 | ||
| AK146675.1 | - | 2 - 3 | 302 | 73 | ||
| BC085127.1 | - | 2 - 3 | 293 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsf1
Genomic Map