Assay Details
| Gene Symbol: Rsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081267.2 | NP_001074736.1 | 9 - 10 | 2921 | 97 | |
| GenBank mRNA | AK032853.1 | - | 9 - 10 | 2897 | 97 | |
| AK146675.1 | - | 9 - 10 | 2931 | 97 | ||
| BC054454.1 | - | 4 - 5 | 412 | 97 | ||
| BC085127.1 | - | 8 - 9 | 3850 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsf1
Genomic Map