Assay Details
| Gene Symbol: Tcerg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183289.3 | NP_899112.2 | 6 - 7 | 1123 | 65 | |
| GenBank mRNA | BC107234.1 | - | 6 - 7 | 738 | 65 | |
| BC107235.1 | - | 6 - 7 | 738 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcerg1l
Genomic Map