Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700015051.mm.1 |
TC0700001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015051.mm.1 |
NM_011247.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015051.mm.1 |
NM_011247.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015053.mm.1 |
TC0700001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015053.mm.1 |
NM_011247.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015053.mm.1 |
NM_011247.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015052.mm.1 |
TC0700001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015052.mm.1 |
NM_011247.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015052.mm.1 |
NM_011247.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015050.mm.1 |
TC0700001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015050.mm.1 |
NM_011247.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015050.mm.1 |
NM_011247.2 |
Refseq |