Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600003187.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003187.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003187.mm.1 |
NM_001286664.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003187.mm.1 |
NM_028358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003187.mm.1 |
NM_028358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003187.mm.1 |
NM_212468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003187.mm.1 |
NM_212468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003184.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003184.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003184.mm.1 |
NM_001286664.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003184.mm.1 |
NM_028358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003184.mm.1 |
NM_028358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003184.mm.1 |
NM_212468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003184.mm.1 |
NM_212468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003183.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003183.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003183.mm.1 |
NM_001286664.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003186.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003186.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003186.mm.1 |
NM_001286664.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003186.mm.1 |
NM_028358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003186.mm.1 |
NM_028358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003186.mm.1 |
NM_212468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003186.mm.1 |
NM_212468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003181.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003181.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003181.mm.1 |
NM_001286664.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003188.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003188.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003188.mm.1 |
NM_001286664.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003188.mm.1 |
NM_028358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003188.mm.1 |
NM_028358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003188.mm.1 |
NM_212468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003188.mm.1 |
NM_212468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003190.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003190.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003190.mm.1 |
NM_001286664.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003190.mm.1 |
NM_028358.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003190.mm.1 |
NM_028358.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003190.mm.1 |
NM_212468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003190.mm.1 |
NM_212468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003180.mm.1 |
TC0600000410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003180.mm.1 |
NM_001286663.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003180.mm.1 |
NM_001286664.1 |
Refseq |