Assay Details
| Gene Symbol: Ap1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290378.1 | NP_001277307.1 | 2 - 3 | 439 | 79 | |
| NM_001290379.1 | NP_001277308.1 | 2 - 3 | 439 | 79 | ||
| NM_026887.3 | NP_081163.2 | 2 - 3 | 439 | 79 | ||
| GenBank mRNA | AK005223.1 | - | 2 - 3 | 303 | 79 | |
| AK043776.1 | - | 2 - 3 | 260 | 79 | ||
| AK045558.1 | - | 2 - 3 | 440 | 79 | ||
| AK047724.1 | - | 2 - 3 | 316 | 79 | ||
| AK053941.1 | - | 2 - 3 | 312 | 79 | ||
| AK167690.1 | - | 2 - 3 | 314 | 79 | ||
| BC046964.1 | - | 2 - 3 | 296 | 6485320 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap1s2
Genomic Map