Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700006383.mm.1 |
NM_001163770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006383.mm.1 |
NM_001163770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006383.mm.1 |
NM_033613.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006383.mm.1 |
NM_033613.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006383.mm.1 |
TC1700000646.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006384.mm.1 |
NM_001163770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006384.mm.1 |
NM_001163770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006384.mm.1 |
NM_033613.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006384.mm.1 |
NM_033613.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006384.mm.1 |
TC1700000646.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006381.mm.1 |
NM_001163770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006381.mm.1 |
NM_001163770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006381.mm.1 |
NM_033613.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006381.mm.1 |
NM_033613.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006381.mm.1 |
TC1700000646.mm.1 |
MTA TC |