Assay Details
| Gene Symbol: Cfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142706.1 | NP_001136178.1 | 4 - 5 | 1056 | 75 | |
| NM_008198.2 | NP_032224.2 | 4 - 5 | 1056 | 75 | ||
| GenBank mRNA | AK098094.1 | - | 4 - 5 | 716 | 75 | |
| AK149531.1 | - | 4 - 5 | 689 | 75 | ||
| AK149573.1 | - | 4 - 5 | 715 | 75 | ||
| AK151246.1 | - | 4 - 5 | 790 | 75 | ||
| BC005451.1 | - | 4 - 5 | 690 | 3591669 | 75 | |
| M57890.1 | - | 4 - 5 | 759 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cfb
Genomic Map