Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700018418.mm.1 |
NM_001146215.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018418.mm.1 |
NM_013600.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018418.mm.1 |
TC1700001940.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018423.mm.1 |
NM_001146215.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018423.mm.1 |
NM_013600.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018423.mm.1 |
TC1700001940.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018425.mm.1 |
NM_013600.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018425.mm.1 |
TC1700001940.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018419.mm.1 |
NM_001146215.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018419.mm.1 |
NM_013600.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018419.mm.1 |
TC1700001940.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018424.mm.1 |
NM_001146215.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018424.mm.1 |
NM_013600.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018424.mm.1 |
TC1700001940.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018420.mm.1 |
NM_001146215.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018420.mm.1 |
NM_013600.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018420.mm.1 |
TC1700001940.mm.1 |
MTA TC |