Assay Details
Gene Symbol: Neu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010893.3 | NP_035023.3 | 4 - 5 | 929 | 64 | |
GenBank mRNA | AK077444.1 | - | 3 - 4 | 2044 | 64 | |
AK145680.1 | - | 4 - 5 | 823 | 64 | ||
AK148229.1 | - | 2 - 3 | 64 | |||
BC004666.1 | - | 4 - 5 | 803 | 3498197 | 64 | |
U93702.1 | - | 4 - 5 | 805 | 64 | ||
Y11412.1 | - | 4 - 5 | 808 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
