Assay Details
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021882.4 | NP_068682.2 | 6 - 7 | 1280 | 72 | |
GenBank mRNA | AF119092.1 | - | 1 - 2 | 157 | 72 | |
AK012808.1 | - | 6 - 7 | 1281 | 72 | ||
AK131956.1 | - | 6 - 7 | 1282 | 72 | ||
AK178741.1 | - | 1 - 2 | 147 | 72 | ||
AK197730.1 | - | 1 - 2 | 147 | 72 | ||
BC075667.1 | - | 6 - 7 | 1265 | 72 | ||
BC082555.1 | - | 6 - 7 | 1266 | 30541702 | 72 | |
U14133.1 | - | 6 - 7 | 1255 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
