Assay Details
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016811.2 | NP_058091.2 | 5 - 6 | 685 | 67 | |
| GenBank mRNA | AF085219.1 | - | 4 - 5 | 371 | 67 | |
| BC006713.1 | - | 5 - 6 | 685 | 3584592 | 67 | |
| JN832677.1 | - | 6 - 7 | 717 | 67 | ||
| JN832678.1 | - | 5 - 6 | 704 | 67 | ||
| JN832679.1 | - | 5 - 6 | 704 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgka
Genomic Map