Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600017185.mm.1 |
TC0600002178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017185.mm.1 |
NM_029930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017185.mm.1 |
NM_029930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017182.mm.1 |
TC0600002178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017182.mm.1 |
NM_029930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017182.mm.1 |
NM_029930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017184.mm.1 |
TC0600002178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017184.mm.1 |
NM_029930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017184.mm.1 |
NM_029930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017188.mm.1 |
TC0600002178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017188.mm.1 |
NM_029930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017188.mm.1 |
NM_029930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017181.mm.1 |
TC0600002178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017181.mm.1 |
NM_029930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017181.mm.1 |
NM_029930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017190.mm.1 |
TC0600002178.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017190.mm.1 |
NM_029930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017190.mm.1 |
NM_029930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017191.mm.1 |
TC0600002178.mm.1 |
MTA TC |