Assay Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013491.2 | NP_038519.1 | 10 - 11 | 1250 | 72 | |
| GenBank mRNA | AK036735.1 | - | 10 - 11 | 1252 | 72 | |
| AK161489.1 | - | 3 - 4 | 215 | 72 | ||
| AY046403.1 | - | 10 - 11 | 1075 | 72 | ||
| AY046404.1 | - | 11 - 12 | 1154 | 72 | ||
| BC114336.1 | - | 10 - 11 | 1215 | 40048966 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcn1
Genomic Map