Assay Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013491.2 | NP_038519.1 | 7 - 8 | 940 | 86 | |
| GenBank mRNA | AK036735.1 | - | 7 - 8 | 942 | 86 | |
| AY046403.1 | - | 7 - 8 | 765 | 86 | ||
| AY046404.1 | - | 8 - 9 | 844 | 86 | ||
| BC114336.1 | - | 7 - 8 | 905 | 40048966 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clcn1
Genomic Map