Assay Details
Gene Symbol: Galnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144908.3 | NP_659157.1 | 5 - 6 | 886 | 73 | |
GenBank mRNA | AF495528.1 | - | 4 - 5 | 758 | 73 | |
AK088014.1 | - | 5 - 6 | 875 | 73 | ||
AK161524.1 | - | 5 - 6 | 796 | 73 | ||
BC011428.1 | - | 5 - 6 | 862 | 2631696 | 73 | |
BC021504.1 | - | 5 - 6 | 887 | 5346678 | 73 | |
BC031150.1 | - | 4 - 5 | 784 | 73 | ||
BC034184.1 | - | 4 - 5 | 786 | 4974216 | 73 | |
BC034185.1 | - | 4 - 5 | 838 | 4974695 | 73 | |
BC036143.1 | - | 4 - 5 | 787 | 4974645 | 73 | |
BC036145.1 | - | 4 - 5 | 780 | 4975474 | 73 | |
Y12435.1 | - | 4 - 5 | 709 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
