Assay Details
Gene Symbol: Nub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018166.1 | - | 2 - 3 | 165 | 151 | |
AK134581.1 | - | 2 - 3 | 168 | 151 | ||
BC005640.1 | - | 2 - 3 | 153 | 151 | ||
BC037451.1 | - | 1 - 2 | 143 | 4018648 | 151 | |
U27462.1 | - | 2 - 3 | 361 | 151 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK134581.1, BC005640.1, BC037451.1, U27462.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
