Assay Details
| Gene Symbol: Ptpn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011203.2 | NP_035333.2 | 8 - 9 | 742 | 154 | |
| GenBank mRNA | AK154107.1 | - | 8 - 9 | 834 | 154 | |
| BC051980.1 | - | 8 - 9 | 742 | 6402545 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn12
Genomic Map