Assay Details
| Gene Symbol: Pmpcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028431.2 | NP_082707.1 | 1 - 2 | 118 | 111 | |
| GenBank mRNA | AK013995.1 | - | 1 - 2 | 119 | 111 | |
| AK152673.1 | - | 1 - 2 | 119 | 111 | ||
| BC060662.1 | - | 1 - 2 | 111 | 111 | ||
| BC065095.1 | - | 1 - 2 | 111 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pmpcb
Genomic Map