Assay Details
| Gene Symbol: Cd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159555.1 | NP_001153027.1 | 7 - 8 | 1160 | 69 | |
| NM_001159556.1 | NP_001153028.1 | 7 - 8 | 1020 | 69 | ||
| NM_001159557.1 | NP_001153029.1 | 8 - 9 | 1071 | 69 | ||
| NM_001159558.1 | NP_001153030.1 | 8 - 9 | 1187 | 69 | ||
| NM_007643.4 | NP_031669.3 | 7 - 8 | 1163 | 69 | ||
| GenBank mRNA | AK052825.1 | - | 7 - 8 | 964 | 69 | |
| AK128908.1 | - | 8 - 9 | 964 | 69 | ||
| AK150648.1 | - | 7 - 8 | 984 | 69 | ||
| AK150865.1 | - | 3 - 4 | 358 | 69 | ||
| AK151355.1 | - | 7 - 8 | 999 | 69 | ||
| AK152642.1 | - | 2 - 3 | 155 | 69 | ||
| AK152904.1 | - | 5 - 6 | 670 | 69 | ||
| AK172162.1 | - | 8 - 9 | 1009 | 69 | ||
| BC010262.1 | - | 7 - 8 | 972 | 3481681 | 69 | |
| L23108.1 | - | 7 - 8 | 981 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd36
Genomic Map