Assay Details
| Gene Symbol: Riok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025934.2 | NP_080210.1 | 5 - 6 | 867 | 138 | |
| GenBank mRNA | AK008385.1 | - | 5 - 6 | 626 | 138 | |
| AK008451.1 | - | 5 - 6 | 627 | 138 | ||
| AK049232.1 | - | 5 - 6 | 611 | 138 | ||
| AK090011.1 | - | 5 - 6 | 630 | 138 | ||
| BC010781.1 | - | 5 - 6 | 608 | 4216633 | 138 | |
| BC080771.1 | - | 5 - 6 | 778 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Riok2
Genomic Map