Assay Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135727.2 | NP_001129199.1 | 11 - 12 | 1559 | 84 | |
| NM_001135728.2 | NP_001129200.1 | 6 - 7 | 561 | 84 | ||
| NM_021389.6 | NP_067364.2 | 10 - 11 | 1124 | 84 | ||
| NR_110969.1 | 10 - 11 | 1124 | 84 | |||
| GenBank mRNA | AK004636.1 | - | 10 - 11 | 1124 | 84 | |
| AK028299.1 | - | 6 - 7 | 565 | 84 | ||
| AK030839.1 | - | 10 - 11 | 1125 | 84 | ||
| AK038540.1 | - | 13 - 14 | 1423 | 84 | ||
| AK049182.1 | - | 11 - 12 | 1560 | 84 | ||
| AK080930.1 | - | 11 - 12 | 1424 | 84 | ||
| AK161735.1 | - | 6 - 7 | 642 | 84 | ||
| AK172022.1 | - | 11 - 12 | 1540 | 84 | ||
| AK213880.1 | - | 2 - 3 | 125 | 84 | ||
| BC034646.1 | - | 10 - 11 | 1128 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3kbp1
Genomic Map