Assay Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172815.3 | NP_766403.1 | 2 - 3 | 963 | 71 | |
| GenBank mRNA | AK049891.1 | - | 2 - 3 | 714 | 71 | |
| AK052457.1 | - | 1 - 2 | 1087 | 71 | ||
| AK087485.1 | - | 2 - 3 | 964 | 71 | ||
| BC052844.1 | - | 2 - 3 | 314 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rspo2
Genomic Map