Assay Details
| Gene Symbol: Rspo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172815.3 | NP_766403.1 | 4 - 5 | 1298 | 75 | |
| GenBank mRNA | AK049891.1 | - | 4 - 5 | 1049 | 75 | |
| AK052457.1 | - | 3 - 4 | 1422 | 75 | ||
| AK087485.1 | - | 4 - 5 | 1299 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rspo2
Genomic Map