Assay Details
Gene Symbol: 1700018A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029439.1 | 2 - 3 | 262 | 63 | ||
GenBank mRNA | AK006076.1 | - | 2 - 3 | 255 | 63 | |
AK016091.1 | - | 2 - 3 | 290 | 63 | ||
BC049749.1 | - | 281 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 11377 | Gm11377 | OTTMUSG00000000685 | -12.2kb | Anti-sense | Chr. 13 - 31564117 - 31567391 on NCBI Build GRCm38 |
Downstream | forkhead box F2 | Foxf2 |
FREAC2, Fkh20, LUN |
45.7kb | Anti-sense | Chr. 13 - 31625816 - 31631406 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
