Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300010547.mm.1 |
NM_001164058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010547.mm.1 |
NM_001164058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010547.mm.1 |
NM_008930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010547.mm.1 |
NM_008930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010547.mm.1 |
TC1300001695.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010548.mm.1 |
NM_001164058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010548.mm.1 |
NM_001164058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010548.mm.1 |
NM_008930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010548.mm.1 |
NM_008930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010548.mm.1 |
TC1300001695.mm.1 |
MTA TC |