Assay Details
| Gene Symbol: Cdkal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018346.1 | - | 3 - 4 | 298 | 97 | |
| AK041864.1 | - | 3 - 4 | 294 | 97 | ||
| AK045506.1 | - | 3 - 4 | 318 | 97 | ||
| AK082629.1 | - | 3 - 4 | 301 | 97 | ||
| AK158546.1 | - | 3 - 4 | 291 | 97 | ||
| AK169761.1 | - | 3 - 4 | 298 | 97 | ||
| BC016073.1 | - | 3 - 4 | 242 | 4482130 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdkal1
Genomic Map