| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600016082.mm.1 |
TC0600002045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016082.mm.1 |
NM_009658.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016082.mm.1 |
NM_009658.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016082.mm.1 |
NR_037965.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016086.mm.1 |
TC0600002045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016086.mm.1 |
NM_009658.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016086.mm.1 |
NM_009658.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016086.mm.1 |
NR_037965.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016083.mm.1 |
TC0600002045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016083.mm.1 |
NM_009658.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016083.mm.1 |
NM_009658.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016083.mm.1 |
NR_037965.1 |
Refseq |