Assay Details
| Gene Symbol: Ggps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010282.2 | NP_034412.1 | 3 - 4 | 358 | 79 | |
| GenBank mRNA | AB016044.1 | - | 3 - 4 | 246 | 79 | |
| AK011980.1 | - | 4 - 5 | 503 | 79 | ||
| AK132907.1 | - | 3 - 4 | 351 | 79 | ||
| BC006798.1 | - | 3 - 4 | 343 | 2651229 | 79 | |
| BC069913.1 | - | 3 - 4 | 358 | 6469191 | 79 | |
More Information
Set Membership: |
|

Ggps1
Genomic Map