Assay Details
| Gene Symbol: Prss16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019429.2 | NP_062302.1 | 9 - 10 | 1190 | 79 | |
| GenBank mRNA | AJ131775.1 | - | 9 - 10 | 1190 | 79 | |
| AK040376.1 | - | 2 - 3 | 573 | 79 | ||
| AK088019.1 | - | 9 - 10 | 1218 | 79 | ||
| BC125481.1 | - | 9 - 10 | 1188 | 40129996 | 79 | |
| BC131955.1 | - | 9 - 10 | 1188 | 40130232 | 79 | |
More Information
Set Membership: |
|

Prss16
Genomic Map