Assay Details
Gene Symbol: Gpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008156.2 | NP_032182.2 | 21 - 22 | 2340 | 96 | |
GenBank mRNA | AF050666.1 | - | 21 - 22 | 2279 | 96 | |
AK004957.2 | - | 20 - 21 | 2044 | 96 | ||
AY081194.1 | - | 20 - 21 | 2067 | 96 | ||
AY333960.1 | - | 20 - 21 | 2024 | 96 | ||
BC019146.1 | - | 20 - 21 | 2053 | 5052822 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
