Assay Details
| Gene Symbol: Gpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008156.2 | NP_032182.2 | 5 - 6 | 647 | 80 | |
| GenBank mRNA | AK004957.2 | - | 4 - 5 | 351 | 80 | |
| AY081194.1 | - | 4 - 5 | 374 | 80 | ||
| AY333960.1 | - | 4 - 5 | 331 | 80 | ||
| BC019146.1 | - | 4 - 5 | 360 | 5052822 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpld1
Genomic Map