Assay Details
| Gene Symbol: Ppme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028292.2 | NP_082568.1 | 11 - 12 | 1083 | 119 | |
| GenBank mRNA | AK028261.1 | - | 11 - 12 | 1118 | 119 | |
| AK076992.1 | - | 11 - 12 | 1494 | 119 | ||
| AK146051.1 | - | 11 - 12 | 1065 | 119 | ||
| AK146268.1 | - | 11 - 12 | 1095 | 119 | ||
| AK154876.1 | - | 11 - 12 | 1156 | 119 | ||
| AK208822.1 | - | 2 - 3 | 104 | 119 | ||
| AK216235.1 | - | 2 - 3 | 104 | 119 | ||
| BC014867.1 | - | 11 - 12 | 1083 | 3596696 | 119 | |
| BC029064.1 | - | 11 - 12 | 1071 | 4952806 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppme1
Genomic Map