Assay Details
| Gene Symbol: Ucp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009464.3 | NP_033490.1 | 1 - 2 | 103 | 87 | |
| GenBank mRNA | AB010742.1 | - | 1 - 2 | 103 | 87 | |
| AF032902.1 | - | 1 - 2 | 70 | 87 | ||
| AF053352.1 | - | 1 - 2 | 75 | 87 | ||
| BC055901.1 | - | 1 - 2 | 100 | 87 | ||
| BC098234.1 | - | 1 - 2 | 113 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB010742.1, AF032902.1, AF053352.1, NM_009464.3 transcripts.
Set Membership: |
|

Ucp3
Genomic Map