Assay Details
| Gene Symbol: Mrpl48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198831.2 | NP_942128.2 | 2 - 3 | 198 | 95 | |
| NR_003559.1 | 2 - 3 | 163 | 95 | |||
| GenBank mRNA | AK007653.1 | - | 2 - 3 | 469 | 95 | |
| AK019161.1 | - | 2 - 3 | 160 | 95 | ||
| AK171917.1 | - | 2 - 3 | 171 | 95 | ||
| AK211110.1 | - | 1 - 2 | 60 | 95 | ||
| BC037190.1 | - | 2 - 3 | 153 | 4168034 | 95 | |
| BC116358.1 | - | 1 - 2 | 68 | 40099962 | 95 | |
| BC116359.1 | - | 1 - 2 | 65 | 40099963 | 95 | |
| BC128487.1 | - | 1 - 2 | 68 | 40099964 | 95 | |
| BC128488.1 | - | 1 - 2 | 68 | 40099965 | 95 | |
| BU604260.1 | - | 2 - 3 | 191 | 95 | ||
| CD363582.1 | - | 2 - 3 | 129 | 95 | ||
| DN174444.1 | - | 2 - 3 | 198 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116358.1, BC116359.1, BC128487.1, BC128488.1 transcripts.
Set Membership: |
|

Mrpl48
Genomic Map