Assay Details
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027629.3 | NP_081905.1 | 2 - 3 | 595 | 100 | |
GenBank mRNA | AK039189.1 | - | 2 - 3 | 579 | 100 | |
AK040513.1 | - | 2 - 3 | 515 | 100 | ||
BC020351.1 | - | 2 - 3 | 808 | 100 | ||
BC076571.1 | - | 2 - 3 | 492 | 30356163 | 100 | |
CF531609.1 | - | 2 - 3 | 492 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
