Assay Details
| Gene Symbol: Trim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285870.1 | NP_001272799.1 | 8 - 9 | 1930 | 61 | |
| NM_001285871.1 | NP_001272800.1 | 8 - 9 | 2115 | 61 | ||
| NM_001285873.1 | NP_001272802.1 | 8 - 9 | 1930 | 61 | ||
| NM_018880.3 | NP_061368.1 | 8 - 9 | 1904 | 61 | ||
| GenBank mRNA | AB030912.1 | - | 8 - 9 | 1931 | 61 | |
| AF220019.1 | - | 8 - 9 | 1823 | 61 | ||
| AK019165.1 | - | 8 - 9 | 1865 | 61 | ||
| AK144674.1 | - | 8 - 9 | 1833 | 61 | ||
| AK170029.1 | - | 8 - 9 | 1868 | 61 | ||
| BC003347.1 | - | 8 - 9 | 1944 | 61 | ||
| BC034263.1 | - | 8 - 9 | 1869 | 4503884 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim3
Genomic Map