Assay Details
| Gene Symbol: Slc37a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028123.3 | NP_082399.3 | 10 - 11 | 1186 | 73 | |
| GenBank mRNA | AK012071.1 | - | 2 - 3 | 409 | 73 | |
| AK076213.1 | - | 10 - 11 | 1129 | 73 | ||
| AK077836.1 | - | 10 - 11 | 1129 | 73 | ||
| AK167716.1 | - | 10 - 11 | 1142 | 73 | ||
| BC005744.1 | - | 10 - 11 | 1005 | 73 | ||
| BC099673.1 | - | 11 - 12 | 1123 | 4936456 | 73 | |
| BC100391.1 | - | 11 - 12 | 1220 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc37a3
Genomic Map