Assay Details
Gene Symbol: BC051019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040700.2 | NP_001035790.1 | 3 - 4 | 714 | 106 | |
GenBank mRNA | AK143006.1 | - | 3 - 4 | 402 | 106 | |
BC051019.1 | - | 3 - 4 | 714 | 6390147 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
