Assay Details
Gene Symbol: BC051019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040700.2 | NP_001035790.1 | 4 - 5 | 945 | 101 | |
GenBank mRNA | AK143006.1 | - | 4 - 5 | 633 | 101 | |
BC051019.1 | - | 4 - 5 | 945 | 6390147 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038187 | NM_001040700.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038187 | AK143006.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2038187 | BC051019.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92309 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368875 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810357 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225703 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654485 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887415 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178213 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104949 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120230 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038187 | NM_001040700.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038187 | AK143006.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2038187 | BC051019.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P164255 | NM_001040700.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P164255 | AK143006.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P164255 | BC051019.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164255 | NM_001040700.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164255 | AK143006.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P164255 | BC051019.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293104 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293106 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06475527 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262228 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06293100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25409635 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06030010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06262226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06045853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409626 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06262228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06568530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05980810 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409653 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06190313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06539593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06482002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06293102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06604383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06604384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25409619 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10566795 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566792 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566796 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566797 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566799 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566794 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566798 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419338_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566791 | - | - |
Mouse Genome 430 2.0 Array | 1419338_at | - | - |
Mouse Genome 430 2.0 Array | 1419338_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57355_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57355_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700033685.mm.1 | NM_001040700.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033685.mm.1 | NM_001040700.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033685.mm.1 | TC0700004078.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700033686.mm.1 | NM_001040700.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033686.mm.1 | NM_001040700.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033686.mm.1 | TC0700004078.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700033687.mm.1 | NM_001040700.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033687.mm.1 | NM_001040700.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033687.mm.1 | TC0700004078.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419338_at | - | - |
Mouse Expression Set 430 | 1419338_PM_at | - | - |
Murine Genome U74v2 Set | 102640_PM_at | - | - |
Murine Genome U74v2 Set | 102640_at | - | - |
Murine Genome U74v2 Set | 134057_at | - | - |
Murine Genome U74v2 Set | 134057_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419338_at | - | - |
Mouse Genome 430A 2.0 Array | 1419338_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419338_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419338_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 539987 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_2875039 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2875039 | - | - |
